missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V delta 2 Antibody (15D), Novus Biologicals™
Mouse Monoclonal Antibody
213.00€
Specifications
| Antigen | TCR V delta 2 |
|---|---|
| Clone | 15D |
| Concentration | 0.15 mg/mL |
| Dilution | Western Blot 1:1 - 1:100, Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay |
Description
TCR V delta 2 Monoclonal specifically detects TCR V delta 2 in Human, Drosophila, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| TCR V delta 2 | |
| 0.15 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay | |
| Unconjugated | |
| Mouse | |
| Human, Drosophila, Primate | |
| 6964 | |
| Human TCR V delta 2 | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 15D | |
| Western Blot 1:1 - 1:100, Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional | |
| Monoclonal | |
| Ascites | |
| RUO | |
| T cell receptor delta locus, T-cell antigen receptor, delta polypeptide, T-cell receptor, delta (V,D,J,C), TCRD, TCRD, TRD@, TCRDV1, TRD@ | |
| TRD | |
| IgG1 | |
| Protein A purified | |
| This antibody reacts with Human. Drosophila reactivity reported in scientific literature (PMID: 18802083). Rhesus Monkey reactivity reported in scientific literature (PMID: 20660793). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title