missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nse2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
466.00€
Specifications
| Antigen | Nse2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Nse2 Polyclonal specifically detects Nse2 in Human samples. It is validated for Western Blot.Specifications
| Nse2 | |
| Polyclonal | |
| Rabbit | |
| Q96MF7 | |
| 286053 | |
| Synthetic peptides corresponding to NSMCE2(non-SMC element 2, MMS21 homolog (S. cerevisiae)) The peptide sequence was selected from the middle region of NSMCE2. Peptide sequence KFLALQSKNSDADFQNNEKFVQFKQQLKELKKQCGLQADREADGTEGVDE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| C8orf36, E3 SUMO-protein ligase NSE2, EC 6.3.2, EC 6.3.2.-, FLJ32440, hMMS21, methyl methanesulfonate sensitivity gene 21, MMS21 homolog, MMS21chromosome 8 open reading frame 36, Non-SMC element 2 homolog, non-SMC element 2 homolog (MMS21, S. cerevisiae), non-SMC element 2, MMS21 homolog (S. cerevisiae), Non-structural maintenance of chromosomes element 2 homolog, NSE2 | |
| NSMCE2 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title