missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nse2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56293
This item is not returnable.
View return policy
Description
Nse2 Polyclonal specifically detects Nse2 in Human samples. It is validated for Western Blot.
Specifications
| Nse2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C8orf36, E3 SUMO-protein ligase NSE2, EC 6.3.2, EC 6.3.2.-, FLJ32440, hMMS21, methyl methanesulfonate sensitivity gene 21, MMS21 homolog, MMS21chromosome 8 open reading frame 36, Non-SMC element 2 homolog, non-SMC element 2 homolog (MMS21, S. cerevisiae), non-SMC element 2, MMS21 homolog (S. cerevisiae), Non-structural maintenance of chromosomes element 2 homolog, NSE2 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 286053 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96MF7 | |
| NSMCE2 | |
| Synthetic peptides corresponding to NSMCE2(non-SMC element 2, MMS21 homolog (S. cerevisiae)) The peptide sequence was selected from the middle region of NSMCE2. Peptide sequence KFLALQSKNSDADFQNNEKFVQFKQQLKELKKQCGLQADREADGTEGVDE. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Pig: 100%. | |
| Human, Mouse, Bovine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction