missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPM1 Antibody (FC82291), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
222.00€
Specifications
| Antigen | NPM1 |
|---|---|
| Clone | FC82291 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-4 μg/mL, Immunoblotting 0.5 μg/mL, Immunofluorescence 1:500 |
| Applications | Western Blot |
Description
NPM1 Monoclonal specifically detects NPM1 in Human, Mouse, Rat, Bovine, Canine, Donkey, Hamster, Monkey samples. It is validated for Western Blot, Immunoblotting, Immunofluorescence.Specifications
| NPM1 | |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Mitotic Regulators, Tumor Suppressors | |
| B23, MGC104254, NPMnucleophosmin/nucleoplasmin family, member 1, Nucleolar phosphoprotein B23, Nucleolar protein NO38, nucleophosmin, nucleophosmin (nucleolar phosphoprotein B23, numatrin), numatrin | |
| NPM1 | |
| IgG1 | |
| Protein A or G purified | |
| Detects both the phosphorylated and the unphosphorylated B23 molecule. |
| FC82291 | |
| Western Blot 2-4 μg/mL, Immunoblotting 0.5 μg/mL, Immunofluorescence 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 4869 | |
| rat B23 (nucleophosmin, NPM, numatrin). | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title