missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPM1 Antibody (FC82291), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB600-1030-0.025ml
This item is not returnable.
View return policy
Description
NPM1 Monoclonal specifically detects NPM1 in Human, Mouse, Rat, Bovine, Canine, Donkey, Hamster, Monkey samples. It is validated for Western Blot, Immunoblotting, Immunofluorescence.
Specifications
| NPM1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 2-4 μg/mL, Immunoblotting 0.5 μg/mL, Immunofluorescence 1:500 | |
| NPM1 | |
| rat B23 (nucleophosmin, NPM, numatrin). | |
| 0.025 mL | |
| Cancer, Cell Cycle and Replication, Mitotic Regulators, Tumor Suppressors | |
| 4869 | |
| Human | |
| Purified |
| Western Blot | |
| FC82291 | |
| Unconjugated | |
| B23, MGC104254, NPMnucleophosmin/nucleoplasmin family, member 1, Nucleolar phosphoprotein B23, Nucleolar protein NO38, nucleophosmin, nucleophosmin (nucleolar phosphoprotein B23, numatrin), numatrin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Detects both the phosphorylated and the unphosphorylated B23 molecule. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction