missing translation for 'onlineSavingsMsg'
Learn More
Learn More
nNOS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
186.00€ - 563.00€
Specifications
| Antigen | nNOS |
|---|---|
| Dilution | Western Blot 1:250, Immunohistochemistry 20 ug/ml, Immunohistochemistry-Paraffin 20 ug/ml |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18037619
|
Novus Biologicals
NBP1-97555-0.025MG |
0.025 mg |
186.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18037809
|
Novus Biologicals
NBP1-97555-0.05MG |
0.05 mg |
391.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18040309
|
Novus Biologicals
NBP1-97555-0.2MG |
0.2 mg |
563.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
nNOS Polyclonal specifically detects nNOS in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| nNOS | |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse | |
| bNOS, brain, Constitutive NOS, EC 1.14.13.39, IHPS1, Neuronal NOS, nitric oxide synthase 1 (neuronal), N-NOS, nNOSNOS type I, NOS, Peptidyl-cysteine S-nitrosylase NOS1 | |
| NOS1 | |
| IgG | |
| Protein A purified |
| Western Blot 1:250, Immunohistochemistry 20 ug/ml, Immunohistochemistry-Paraffin 20 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 4842 | |
| Synthetic peptide corresponding to the sequence near the C-terminus of human nNOS. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title