missing translation for 'onlineSavingsMsg'
Learn More
Learn More
nNOS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97555-0.2MG
This item is not returnable.
View return policy
Description
nNOS Polyclonal specifically detects nNOS in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| nNOS | |
| Polyclonal | |
| Western Blot 1:250, Immunohistochemistry 20 ug/ml, Immunohistochemistry-Paraffin 20 ug/ml | |
| bNOS, brain, Constitutive NOS, EC 1.14.13.39, IHPS1, Neuronal NOS, nitric oxide synthase 1 (neuronal), N-NOS, nNOSNOS type I, NOS, Peptidyl-cysteine S-nitrosylase NOS1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 4842 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| NOS1 | |
| Synthetic peptide corresponding to the sequence near the C-terminus of human nNOS. | |
| 0.2 mg | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction