missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX3-2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93813-0.02ml
This item is not returnable.
View return policy
Description
NKX3-2 Polyclonal antibody specifically detects NKX3-2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NKX3-2 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500 | |
| Bagpipe homeobox protein homolog 1, BAPX1bagpipe homeobox homolog 1 (Drosophila), Homeobox protein NK-3 homolog B, homeobox protein Nkx-3.2, MGC138171, NK3 homeobox 2, NKX3.2, NKX3BSMMD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human NKX3-2 (NP_001180.1). MAVRGANTLTSFSIQAILNKKEERGGLAAPEGRPAPGGTAASVAAAPAVCCWRLFGERDAGALGGAEDSLLASPAGTRTAAGRTAESPEGWDSDSALSEENESRRRCADARGASGAGLAGGSLSLGQPVCELAASKDLEEEAAGRSDSEMSASVSGDRSPRTEDDGVGPRGAHVSALCSG | |
| 0.02 mL | |
| Apoptosis, Cell Biology | |
| 579 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction