missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKR-P1D Antibody (4E9) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
188.00€ - 351.00€
Specifications
| Antigen | NKR-P1D |
|---|---|
| Clone | 4E9 |
| Concentration | 1 mg/mL |
| Applications | Functional Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653109
|
Novus Biologicals
NBP2-50531-0.025ml |
0.025 mL |
188.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682538
|
Novus Biologicals
NBP2-50531 |
0.1 mL |
351.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKR-P1D Monoclonal antibody specifically detects NKR-P1D in Mouse samples. It is validated for Functional AssaySpecifications
| NKR-P1D | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| YB2/0 rat plasmacytoma cells transfected with NKR-P1D | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 4E9 | |
| Functional Assay | |
| Unconjugated | |
| Rat | |
| Mouse | |
| AI462337, CD161, Klrb1b, Klrb1c, Klrb1d, Ly55b, Ly55c, ly-55c, Ly55d, Ly59, Ly-59, Nk1, Nk-1, NK1.1, Nk1.2, Nk-1.2, NKRP1, NK-RP1, NKR-P1.9, NKRP140, Nkrp1b, Nkrp1-b, NKR-P1B, Nkrp1c, Nkrp1d, NKR-P1D | |
| IgG2b | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title