Learn More
NKR-P1D Antibody (4E9) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50531-0.025ml
This item is not returnable.
View return policy
Description
NKR-P1D Monoclonal antibody specifically detects NKR-P1D in Mouse samples. It is validated for Functional Assay
Specifications
| NKR-P1D | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Mouse | |
| Purified |
| Functional Assay | |
| 4E9 | |
| Unconjugated | |
| AI462337, CD161, Klrb1b, Klrb1c, Klrb1d, Ly55b, Ly55c, ly-55c, Ly55d, Ly59, Ly-59, Nk1, Nk-1, NK1.1, Nk1.2, Nk-1.2, NKRP1, NK-RP1, NKR-P1.9, NKRP140, Nkrp1b, Nkrp1-b, NKR-P1B, Nkrp1c, Nkrp1d, NKR-P1D | |
| YB2/0 rat plasmacytoma cells transfected with NKR-P1D | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |