missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 3/CHRNA3 Antibody (6D3A10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.00€ - 557.00€
Specifications
| Antigen | Nicotinic Acetylcholine R alpha 3/CHRNA3 |
|---|---|
| Clone | 6D3A10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18275511
|
Novus Biologicals
NBP2-61673 |
0.1 mL |
557.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666788
|
Novus Biologicals
NBP2-61673-0.025ml |
0.025 mL |
217.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nicotinic Acetylcholine R alpha 3/CHRNA3 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 3/CHRNA3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Nicotinic Acetylcholine R alpha 3/CHRNA3 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| 1136 | |
| Purified recombinant fragment of human CHRNA3 (AA: 32-240) expressed in E. Coli. | |
| Primary | |
| 57.5 kDa |
| 6D3A10 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| cholinergic receptor, nicotinic, alpha 3, cholinergic receptor, nicotinic, alpha polypeptide 3, LNCR2, MGC104879, NACHRA3, neuronal acetylcholine receptor subunit alpha-3, neuronal nicotinic acetylcholine receptor, alpha3 subunit, PAOD2 | |
| CHRNA3 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title