missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acetylcholine R alpha 3/CHRNA3 Antibody (6D3A10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-61673-0.025ml

 View more versions of this product

Product Code. 18666788

  • 217.00€ / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Nicotinic Acetylcholine R alpha 3/CHRNA3 Monoclonal specifically detects Nicotinic Acetylcholine R alpha 3/CHRNA3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nicotinic Acetylcholine R alpha 3/CHRNA3
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
cholinergic receptor, nicotinic, alpha 3, cholinergic receptor, nicotinic, alpha polypeptide 3, LNCR2, MGC104879, NACHRA3, neuronal acetylcholine receptor subunit alpha-3, neuronal nicotinic acetylcholine receptor, alpha3 subunit, PAOD2
Mouse
57.5 kDa
0.025 mL
1136
Purified
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
6D3A10
Unconjugated
PBS with 0.05% Sodium Azide
CHRNA3
Purified recombinant fragment of human CHRNA3 (AA: 32-240) expressed in E. Coli.
Protein G purified
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.