missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicastrin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00€
Specifications
| Antigen | Nicastrin |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
Nicastrin Polyclonal specifically detects Nicastrin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Nicastrin | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin | |
| NCSTN | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| NP_056146 | |
| 23385 | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human Nicastrin. The immunogen is located within amino acids 630 - 680 of Nicastrin. Amino Acid Squence: TEYSTWTESRWKDIRAR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title