missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicastrin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77269-0.025mg
This item is not returnable.
View return policy
Description
Nicastrin Polyclonal specifically detects Nicastrin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Nicastrin | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| NP_056146 | |
| NCSTN | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human Nicastrin. The immunogen is located within amino acids 630 - 680 of Nicastrin. Amino Acid Squence: TEYSTWTESRWKDIRAR | |
| 0.025 mg | |
| Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 23385 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction