missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | NHE2 |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18068075
|
Proteintech
25105-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18040526
|
Proteintech
25105-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NHE2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NHE2. This antibody recognizes human, mouse, rat antigen. The NHE2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NHE2 | |
| Western Blot, ELISA | |
| Rabbit | |
| 6549 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 600 μg/mL | |
| Liquid | |
| Human, Mouse, Rat | |
| Na(+)/H(+) exchanger 2, NHE 2, NHE2, NHE2/SLC9A2, SLC9A2, Sodium/hydrogen exchanger 2 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title