Learn More
NHE2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25105-1-AP-20UL
 View more versions of this product
Product Code. 18040526
  • 145.00€ / 20µL
Description
The NHE2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NHE2. This antibody recognizes human, mouse, rat antigen. The NHE2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
NHE2
600 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, ELISA
Na(+)/H(+) exchanger 2, NHE 2, NHE2, NHE2/SLC9A2, SLC9A2, Sodium/hydrogen exchanger 2
20 μL
6549
Store at -20°C. Stable for one year after shipment.