Learn More
NHE2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25105-1-AP-20UL
This item is not returnable.
View return policy
Description
The NHE2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NHE2. This antibody recognizes human, mouse, rat antigen. The NHE2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NHE2 | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| Na(+)/H(+) exchanger 2, NHE 2, NHE2, NHE2/SLC9A2, SLC9A2, Sodium/hydrogen exchanger 2 | |
| 20 μL | |
| 6549 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |