missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeutraKine™HGF Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
149.00€ - 319.00€
Specifications
| Antigen | HGF |
|---|---|
| Clone | 2D2B4 |
| Applications | ELISA, Neutralization |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235588
|
Proteintech
69027-1-IG-25UG |
25 μg |
149.97€ 149.00€ / 25µg Save 0.97€ 0.65% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234653
|
Proteintech
69027-1-IG-100UG |
100 μg |
321.08€ 319.00€ / 100µg Save 2.08€ 0.65% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NeutraKine™HGF antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine™HGF. This antibody recognizes human antigen. The NeutraKine™HGF antibody has been validated for the following applications: ELISA, Neutralization analysis.Specifications
| HGF | |
| ELISA, Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| F-TCF; HGFB; HPTA; SF | |
| IgG1 | |
| Protein G purification | |
| Neu Antibody |
| 2D2B4 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 3082 | |
| RecombinantProtein | |
| Primary | |
| Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title