missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeutraKine™HGF Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 69027-1-IG-25UG
This item is not returnable.
View return policy
Description
The NeutraKine™HGF antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine™HGF. This antibody recognizes human antigen. The NeutraKine™HGF antibody has been validated for the following applications: ELISA, Neutralization analysis.Specifications
| HGF | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3082 | |
| Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
| Liquid |
| ELISA, Neutralization | |
| 2D2B4 | |
| F-TCF; HGFB; HPTA; SF | |
| RecombinantProtein | |
| 25 μg | |
| Primary | |
| Human | |
| Neu Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction