missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOR2 Antibody (1542), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
200.00€
Specifications
| Antigen | NCOR2 |
|---|---|
| Clone | 1542 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 4 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Applications | Western Blot |
Beschreibung
NCOR2 Monoclonal specifically detects NCOR2 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Frozen.Spezifikation
| NCOR2 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Chromatin Research, Transcription Factors and Regulators | |
| CTG repeat protein 26, CTG26nuclear receptor co-repressor 2, FLJ25011, N-CoR2, nuclear receptor corepressor 2, retinoic-acid-receptor-associated corepressor, silencing mediator for retinoid and thyroid hormone receptors, SMAP270, SMRTE, SMRTE-tau, SMRTT3 receptor-associating factor, TNRC14, TRAC, TRAC1 | |
| NCOR2 | |
| IgG1 | |
| Immunogen affinity purified | |
| NCoR2 - CHIP Grade (1542) |
| 1542 | |
| Western Blot 4 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 9612 | |
| Synthetic peptide corresponding to residues C G G A(2016) S A S D P H R E K T Q(2027) of human NCOR2. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts