missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOR2 Antibody (1542), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB120-2781-0.025mg
This item is not returnable.
View return policy
Description
NCOR2 Monoclonal specifically detects NCOR2 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| NCOR2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 4 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| NCOR2 | |
| Synthetic peptide corresponding to residues C G G A(2016) S A S D P H R E K T Q(2027) of human NCOR2. | |
| 0.025 mg | |
| Cancer, Chromatin Research, Transcription Factors and Regulators | |
| 9612 | |
| Human | |
| Purified |
| Western Blot | |
| 1542 | |
| Unconjugated | |
| CTG repeat protein 26, CTG26nuclear receptor co-repressor 2, FLJ25011, N-CoR2, nuclear receptor corepressor 2, retinoic-acid-receptor-associated corepressor, silencing mediator for retinoid and thyroid hormone receptors, SMAP270, SMRTE, SMRTE-tau, SMRTT3 receptor-associating factor, TNRC14, TRAC, TRAC1 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| NCoR2 - CHIP Grade (1542) | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction