Learn More
NANS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38112-100ul
This item is not returnable.
View return policy
Description
NANS Polyclonal antibody specifically detects NANS in Human samples. It is validated for ELISA,Western Blot
Specifications
| NANS | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 2.5.1.56, EC 2.5.1.57, N-acetylneuraminate synthase, N-acetylneuraminate-9-phosphate synthase, N-acetylneuraminic acid phosphate synthase, N-acetylneuraminic acid synthasesialic acid phosphate synthase, SASsialic acid synthase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 260-359 of human NANS (NP_061819.2).,, Sequence:, AELVRSVRLVERALGSPTKQLLPCEMACNEKLGKSVVAKVKIPEGTILTMDMLTVKVGEPKGYPPEDIFNLVGKKVLVTVEEDDTIMEELVDNHGKKIKS | |
| 100 μL | |
| metabolism, Signal Transduction | |
| 54187 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |