missing translation for 'onlineSavingsMsg'
Learn More

NanoLuc™ (Nluc) Luciferase Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

206.00€ - 490.00€

Specifications

Antigen Luciferase
Clone 965853
Dilution Western Blot 2 μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18731757
GREENER_CHOICE
View Documents
R&D Systems
MAB100261-SP
25 μg
206.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18793607
GREENER_CHOICE
View Documents
R&D Systems
MAB100261-100
100 μg
490.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Luciferase Monoclonal specifically detects Luciferase in Multi-Species samples. It is validated for Western Blot.
Specifications

Specifications

Luciferase
Western Blot 2 μg/mL
Monoclonal
Supernatant
Multi-species
ec 1.13.12.7, Fluc, luciferin 4 monooxygenase
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
965853
Western Blot
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Synthetic peptide for NanoLuc™ (Nluc) Luciferase
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects NanoLuc™ (Nluc) Luciferase in direct ELISAs. In Western blots, no cross-reactivity with Large BiT is observed.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.