missing translation for 'onlineSavingsMsg'
Learn More

n-Tetradecanamide, 98%

Product Code. 11415207
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11415207

Brand: Thermo Scientific Alfa Aesar B22610.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 638-58-4 | C14H29NO | 227.392 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-58-4
Molecular Formula C14H29NO
Molecular Weight (g/mol) 227.392
MDL Number MFCD00025533
InChI Key QEALYLRSRQDCRA-UHFFFAOYSA-N
Synonym myristamide, n-tetradecanamide, myristic amide, myristic acid amide, tetradecylamide, unii-a269j8qg0o, myristicamide, tetradecanoic acid amide, chembl88158
PubChem CID 69492
IUPAC Name tetradecanamide
SMILES CCCCCCCCCCCCCC(=O)N

Specifications

Melting Point 105°C to 107°C
Quantity 100 g
Formula Weight 227.39
Percent Purity 98%
Chemical Name or Material n-Tetradecanamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.