CAS RN 638-58-4
IUPAC Name:
tetradecanamide
Synonyms:
tetradecanamide
Molecular Weight (g/mol):
227.39
Molecular Formula:
C14H29NO
InChi Key:
QEALYLRSRQDCRA-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCC(N)=O
1
–
1
of
1
results
1
n-Tetradecanamide, 98%
CAS: 638-58-4 Molecular Formula: C14H29NO Molecular Weight (g/mol): 227.392 MDL Number: MFCD00025533 InChI Key: QEALYLRSRQDCRA-UHFFFAOYSA-N Synonym: myristamide,n-tetradecanamide,myristic amide,myristic acid amide,tetradecylamide,unii-a269j8qg0o,myristicamide,tetradecanoic acid amide,chembl88158 PubChem CID: 69492 IUPAC Name: tetradecanamide SMILES: CCCCCCCCCCCCCC(=O)N