missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-4-methoxybenzamide, 97%, Thermo Scientific™

Product Code. 15415198
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415198 250 mg 250mg
15405198 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415198 Supplier Thermo Scientific Alfa Aesar Supplier No. H59021.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used to synthesis selective mono-fluorination of 1,2- and 1,3-diols.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7465-86-3
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD00086766
InChI Key HCJXEOFLVIFFDG-UHFFFAOYSA-N
Synonym 4-methoxy-n,n-diethylbenzamide, n,n-diethyl-p-anisamide, n,n-diethyl-4-methoxy-benzamide, benzamide,n,n-diethyl-4-methoxy, n,n-diethyl-para-anisamide, n,n-diethyl 4-methoxyphenyl carboxamide
PubChem CID 231459
IUPAC Name N,N-diethyl-4-methoxybenzamide
SMILES CCN(CC)C(=O)C1=CC=C(C=C1)OC

Specifications

Quantity 250 mg
Formula Weight 207.27
Percent Purity 97%
Chemical Name or Material N,N-Diethyl-4-methoxybenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.