accessibility menu, dialog, popup

UserName

CAS RN 7465-86-3

IUPAC Name: N,N-diethyl-4-methoxybenzamide
Synonyms: N,N-diethyl-4-methoxybenzamide
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: HCJXEOFLVIFFDG-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results