Learn More
N-Acetylmorpholine, 98%
Product Code. 11481127
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11481127
Brand: Thermo Scientific Alfa Aesar B24120.22
Request Bulk or Custom Format
CAS: 1696-20-4 | C6H11NO2 | 129.159 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1696-20-4
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00006171
InChI Key KYWXRBNOYGGPIZ-UHFFFAOYSA-N
Synonym 4-acetylmorpholine, n-acetylmorpholine, 1-morpholinoethanone, morpholine, 4-acetyl, n-acetylmorfolin, n-acetylmorfolin czech, ethanone, 1-4-morpholinyl, n-acetyl morpholine, 1-morpholin-4-yl ethan-1-one, acetylmorpholine
PubChem CID 15543
IUPAC Name 1-morpholin-4-ylethanone
SMILES CC(=O)N1CCOCC1
Specifications
Melting Point 9°C to 10°C
Density 1.116
Boiling Point 245°C to 246°C
Flash Point >110°C (230°F)
Refractive Index 1.4845
Quantity 100 g
Beilstein 111971
Formula Weight 129.16
Percent Purity 98%
Chemical Name or Material N-Acetylmorpholine