CAS RN 1696-20-4
CAS RN 1696-20-4
IUPAC Name:
1-(morpholin-4-yl)ethan-1-one
Synonyms:
morpholine, 4-acetyl-
Molecular Weight (g/mol):
129.16
Molecular Formula:
C6H11NO2
InChi Key:
KYWXRBNOYGGPIZ-UHFFFAOYSA-N
SMILES:
CC(=O)N1CCOCC1
1
–
1
of
1
results
1
N-Acetylmorpholine, 98%
CAS: 1696-20-4 Molecular Formula: C6H11NO2 Molecular Weight (g/mol): 129.159 MDL Number: MFCD00006171 InChI Key: KYWXRBNOYGGPIZ-UHFFFAOYSA-N Synonym: 4-acetylmorpholine,n-acetylmorpholine,1-morpholinoethanone,morpholine, 4-acetyl,n-acetylmorfolin,n-acetylmorfolin czech,ethanone, 1-4-morpholinyl,n-acetyl morpholine,1-morpholin-4-yl ethan-1-one,acetylmorpholine PubChem CID: 15543 IUPAC Name: 1-morpholin-4-ylethanone SMILES: CC(=O)N1CCOCC1