missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYF6 Antibody (5D7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00€ - 552.00€
Specifications
| Antigen | MYF6 |
|---|---|
| Clone | 5D7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18108037
|
Novus Biologicals
NBP2-37479 |
0.1 mL |
552.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633026
|
Novus Biologicals
NBP2-37479-0.025ml |
0.025 mL |
203.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYF6 Monoclonal specifically detects MYF6 in Human samples. It is validated for Western Blot, ELISA.Specifications
| MYF6 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 4618 | |
| Purified recombinant fragment of human MYF6 (AA: 85-150) expressed in E. Coli. | |
| Primary | |
| 27 kDa |
| 5D7 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Human | |
| bHLHc4BHLHC4, Class C basic helix-loop-helix protein 4, MRF4, MRF4myogenic factor 6, Muscle-specific regulatory factor 4, Myf-6, myogenic factor 6 (herculin) | |
| MYF6 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title