missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYF6 Antibody (5D7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37479-0.025ml
This item is not returnable.
View return policy
Description
MYF6 Monoclonal specifically detects MYF6 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| MYF6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| bHLHc4BHLHC4, Class C basic helix-loop-helix protein 4, MRF4, MRF4myogenic factor 6, Muscle-specific regulatory factor 4, Myf-6, myogenic factor 6 (herculin) | |
| Mouse | |
| 27 kDa | |
| 0.025 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, ELISA | |
| 5D7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| MYF6 | |
| Purified recombinant fragment of human MYF6 (AA: 85-150) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 4618 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction