missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium Tuberculosis L-Alanine Dehydrogenase Antibody (1C3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
407.00€
Specifications
| Antigen | Mycobacterium Tuberculosis L-Alanine Dehydrogenase |
|---|---|
| Clone | 1C3 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
Description
Mycobacterium Tuberculosis L-Alanine Dehydrogenase Monoclonal specifically detects Mycobacterium Tuberculosis L-Alanine Dehydrogenase in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
| Mycobacterium Tuberculosis L-Alanine Dehydrogenase | |
| 1.0 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| Culture filtrat from Mycobacterium tuberculosis H37Rv | |
| Primary | |
| Store at 4C. Do not freeze. |
| 1C3 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 40 kDa antigen, Alanine dehydrogenase, EC 1.4.1.1, MT2850, MTV002.45, Rv2780, TB43 | |
| IgG1 κ | |
| Protein A or G purified | |
| Specific for a 40 kDa band from short term culture of Mycobacterium tuberculosis in Western blotting after SDS-PAGE. The 40 kDa band was identified as the protein L-alanine dehydrogenase (Ald). Interspecies analyses with HBT 10 showed reacts with M. tuberculosis and M. marinum but not M. bovis BCG. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title