missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium Tuberculosis L-Alanine Dehydrogenase Antibody (1C3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05061
This item is not returnable.
View return policy
Description
Mycobacterium Tuberculosis L-Alanine Dehydrogenase Monoclonal specifically detects Mycobacterium Tuberculosis L-Alanine Dehydrogenase in Bacteria samples. It is validated for Western Blot, ELISA.
Specifications
| Mycobacterium Tuberculosis L-Alanine Dehydrogenase | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Specific for a 40 kDa band from short term culture of Mycobacterium tuberculosis in Western blotting after SDS-PAGE. The 40 kDa band was identified as the protein L-alanine dehydrogenase (Ald). Interspecies analyses with HBT 10 showed reacts with M. tuberculosis and M. marinum but not M. bovis BCG. | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 1C3 | |
| Unconjugated | |
| 40 kDa antigen, Alanine dehydrogenase, EC 1.4.1.1, MT2850, MTV002.45, Rv2780, TB43 | |
| Culture filtrat from Mycobacterium tuberculosis H37Rv | |
| 100 μg | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction