missing translation for 'onlineSavingsMsg'
Learn More

MUC5AC Antibody (7E9D8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

205.00€ - 552.00€

Specifications

Antigen Mucin 5AC
Clone 7E9D8
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18270265
View Documents
Novus Biologicals
NBP2-61875
0.1 mL
552.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18622669
View Documents
Novus Biologicals
NBP2-61875-0.025ml
0.025 mL
205.00€
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MUC5AC Monoclonal specifically detects MUC5AC in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

Mucin 5AC
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Unconjugated
Mouse
gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin
MUC5AC
IgG2b
Protein G purified
7E9D8
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Monoclonal
Purified
Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction
4586
Purified recombinant fragment of human MUC5AC (AA: 5528-5627) expressed in E. Coli.
Primary
7.5 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.