missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC5AC Antibody (7E9D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61875-0.025ml
This item is not returnable.
View return policy
Description
MUC5AC Monoclonal specifically detects MUC5AC in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Mucin 5AC | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin | |
| Mouse | |
| 7.5 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 7E9D8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| MUC5AC | |
| Purified recombinant fragment of human MUC5AC (AA: 5528-5627) expressed in E. Coli. | |
| Protein G purified | |
| Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction | |
| 4586 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction