missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cystatin B Antibody, R&D Systems™

Rat Monoclonal Antibody
176.00€ - 496.00€
Specifications
| Antigen | Cystatin B/Stefin B |
|---|---|
| Clone | 227807 |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18779058
![]() |
R&D Systems
MAB14091-SP |
25 μg |
176.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18182441
|
R&D Systems
MAB14091 |
500 μg |
496.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cystatin B/Stefin B Monoclonal specifically detects Cystatin B/Stefin B in Mouse samples. It is validated for Immunohistochemistry.Specifications
| Cystatin B/Stefin B | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, CSTB, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B | |
| CSTB | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 227807 | |
| Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q62426 | |
| 1476 | |
| E. coli-derived recombinant mouse Cystatin B Met2-Phe98 Accession # Q62426 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Cystatin B in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Cystatin B is observed and no cross-reactivity with rhCystatin A, C, D, E/M, F, S, SA, rhFetuin A, rhFetuin B, rhHPRG, or rhKininogen is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title