missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cystatin B Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB14091-SP
This item is not returnable.
View return policy
Description
Cystatin B/Stefin B Monoclonal specifically detects Cystatin B/Stefin B in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| Cystatin B/Stefin B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, CSTB, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1476 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry | |
| 227807 | |
| Immunohistochemistry 8-25 ug/mL | |
| Q62426 | |
| CSTB | |
| E. coli-derived recombinant mouse Cystatin B Met2-Phe98 Accession # Q62426 | |
| 25 μg | |
| Primary | |
| Detects mouse Cystatin B in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Cystatin B is observed and no cross-reactivity with rhCystatin A, C, D, E/M, F, S, SA, rhFetuin A, rhFetuin B, rhHPRG, or rhKininogen is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction