missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylmalonyl Coenzyme A mutase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00€ - 529.00€
Specifications
| Antigen | Methylmalonyl Coenzyme A mutase |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18471522
|
Novus Biologicals
NBP2-34024-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18170603
|
Novus Biologicals
NBP2-34024 |
0.1 mL |
529.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methylmalonyl Coenzyme A mutase Polyclonal specifically detects Methylmalonyl Coenzyme A mutase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Methylmalonyl Coenzyme A mutase | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P22033 | |
| 4594 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SVSMTMNGAVIPVLANFIVTGEEQGVPKEKLTGTIQNDILKEFMVRNTYIFPPEPSMKIIADIFEYTAKHMPKFNSISISGYHMQEAGADAILELAYTLADGLEYSRTGLQAGLTIDEFAPRLSFFWGIG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 5.4.99.2, MCMmethylmalonyl Coenzyme A mutase, methylmalonyl CoA mutase, Methylmalonyl-CoA isomerase, methylmalonyl-CoA mutase, mitochondrial | |
| MMUT | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title