missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl pyridazine-4-carboxylate, 96%

Product Code. 11397299
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11397299

Brand: Alfa Aesar™ H27449.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy
Applications
It is used to produce 4-Pyridazincarbohydroxamsaeure by heating and requires a reagent hydroxylammoniumchloride and Na2CO3. Meanwhile, it needs solvent methanol.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34231-77-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD09953488
InChI Key IJFLWNYKGMQQOW-UHFFFAOYSA-N
Synonym pyridazine-4-carboxylic acid methyl ester, 4-pyridazinecarboxylic acid, methyl ester, acmc-1ajnn, methyl 4-pyridazinecarboxylate, methylpyridazine-4-carboxylate, methyl pyridazine-4-carboxylate, 4-pyridazinecarboxylic acid methyl ester
PubChem CID 12439252
IUPAC Name methyl pyridazine-4-carboxylate
SMILES COC(=O)C1=CN=NC=C1

Specifications

Melting Point 64°C to 67°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 138.13
Percent Purity 96%
Chemical Name or Material Methyl pyridazine-4-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.