accessibility menu, dialog, popup

UserName

CAS RN 34231-77-1

CH3OONN

CAS RN 34231-77-1

IUPAC Name: methyl pyridazine-4-carboxylate
Synonyms: methyl pyridazine-4-carboxylate
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: IJFLWNYKGMQQOW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results