missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MEIS1 Antibody (SR3560), Novus Biologicals™
Rabbit Monoclonal Antibody
316.00€ - 382.00€
Specifications
| Antigen | MEIS1 |
|---|---|
| Clone | SR3560 |
| Dilution | Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunoprecipitation 1:50 |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041641
|
Novus Biologicals™
NBP3-27780-50ul |
50 μL |
316.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041615
|
Novus Biologicals™
NBP3-27780-100ul |
100 μL |
382.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEIS1 Monoclonal antibody specifically detects MEIS1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunoprecipitation,Western BlotSpecifications
| MEIS1 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 4211 | |
| IgG | |
| Affinity purified |
| SR3560 | |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| homeobox protein Meis1, leukemogenic homolog protein, Meis homeobox 1, MGC43380, myeloid ecotropic viral integration site 1 homolog, myeloid ecotropic viral integration site 1 homolog (mouse) | |
| A synthesized peptide derived from human MEIS1 (Uniprot #: O00470) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title