missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MEIS1 Antibody (SR3560), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27780-50ul
This item is not returnable.
View return policy
Description
MEIS1 Monoclonal antibody specifically detects MEIS1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot
Specifications
| MEIS1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4211 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| SR3560 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunoprecipitation 1:50 | |
| homeobox protein Meis1, leukemogenic homolog protein, Meis homeobox 1, MGC43380, myeloid ecotropic viral integration site 1 homolog, myeloid ecotropic viral integration site 1 homolog (mouse) | |
| A synthesized peptide derived from human MEIS1 (Uniprot #: O00470) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction