missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM2 Antibody (2B3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37566-0.025ml
This item is not returnable.
View return policy
Description
MCM2 Monoclonal specifically detects MCM2 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MCM2 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| MCM2 | |
| Purified recombinant fragment of human MCM2 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate, Monkey | |
| Ascites |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2B3 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| BM28MGC10606, CCNL 1, CCNL1, CDCL1mitotin, EC 3.6.4.12, KIAA0030MITOTIN, MCM2 minichromosome maintenance deficient 2, mitotin (S. cerevisiae), minichromosome maintenance complex component 2, minichromosome maintenance deficient (S. cerevisiae) 2 (mitotin), minichromosome maintenance deficient 2 (mitotin) | |
| Mouse | |
| 125 kDa | |
| 0.025 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
| 4171 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction