missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM2 Antibody (2B3), Novus Biologicals™
Mouse Monoclonal Antibody
203.00€ - 552.00€
Specifications
| Antigen | MCM2 |
|---|---|
| Clone | 2B3 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629895
|
Novus Biologicals
NBP2-37566-0.025ml |
0.025 mL |
203.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18121819
|
Novus Biologicals
NBP2-37566 |
0.1 mL |
552.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCM2 Monoclonal specifically detects MCM2 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MCM2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat, Primate, Monkey | |
| BM28MGC10606, CCNL 1, CCNL1, CDCL1mitotin, EC 3.6.4.12, KIAA0030MITOTIN, MCM2 minichromosome maintenance deficient 2, mitotin (S. cerevisiae), minichromosome maintenance complex component 2, minichromosome maintenance deficient (S. cerevisiae) 2 (mitotin), minichromosome maintenance deficient 2 (mitotin) | |
| MCM2 | |
| IgG1 | |
| Unpurified |
| 2B3 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
| Ascites with 0.03% Sodium Azide | |
| 4171 | |
| Purified recombinant fragment of human MCM2 expressed in E. Coli. | |
| Primary | |
| 125 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title