missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Matriptase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
201.60€ - 552.30€
Specifications
| Antigen | Matriptase 2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230054
|
Novus Biologicals
NBP3-35158-100ul |
100 μL |
584.00€ 552.30€ / 100µL Save 31.70€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228036
|
Novus Biologicals
NBP3-35158-20ul |
20 μL |
214.00€ 201.60€ / 20µL Save 12.40€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Matriptase 2 Polyclonal antibody specifically detects Matriptase 2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| Matriptase 2 | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis | |
| PBS (pH 7.3), 50% glycerol | |
| 164656 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.4.21, EC 3.4.21.-, FLJ30744, IRIDA, Matriptase-2, membrane-bound mosaic serine proteinase matriptase-2, transmembrane protease serine 6, transmembrane protease, serine 6, type II transmembrane serine protease 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 77-220 of human Matriptase 2 (NP_705837.1).,, Sequence:, GYKAEVMVSQVYSGSLRVLNRHFSQDLTRRESSAFRSETAKAQKMLKELITSTRLGTYYNSSSVYSFGEGPLTCFFWFILQIPEHRRLMLSPEVVQALLVEELLSTVNSSAAVPYRAEYEVDPEGLVILEASVKDIAALNSTLG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title