missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Matriptase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35158-20ul
This item is not returnable.
View return policy
Description
Matriptase 2 Polyclonal antibody specifically detects Matriptase 2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Matriptase 2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| EC 3.4.21, EC 3.4.21.-, FLJ30744, IRIDA, Matriptase-2, membrane-bound mosaic serine proteinase matriptase-2, transmembrane protease serine 6, transmembrane protease, serine 6, type II transmembrane serine protease 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 77-220 of human Matriptase 2 (NP_705837.1).,, Sequence:, GYKAEVMVSQVYSGSLRVLNRHFSQDLTRRESSAFRSETAKAQKMLKELITSTRLGTYYNSSSVYSFGEGPLTCFFWFILQIPEHRRLMLSPEVVQALLVEELLSTVNSSAAVPYRAEYEVDPEGLVILEASVKDIAALNSTLG | |
| 20 μL | |
| Angiogenesis | |
| 164656 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction