Learn More
Eppendorf™ Mastercycler X50s 96-Well Silver Block Thermal Cycler PROMO
GREENER_CHOICE
Product Code. 15896721 Shop All Eppendorf Products
Change view
Click to view available options
Dimensions:
27.5 x 43 x 33 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions Unit Size
15896721 27.5 x 43 x 33 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15896721 Supplier Eppendorf™ Supplier No. 6311000010
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 28-12-2026
Boost your PCR speed – save with the Mastercycler™ X50s and X50i Silver Block bundle
Mastercycler™ X50 Sliver block Bundle
  • One unit of X50s and one unit of X50i bundle
  • Fast heating rate: max. 10°C/s
  • High block temperature homogeneity (±0.2°C at 20 to 72°C) and control accuracy (±0.15°C)
  • Low noise level (33.6 dB(A)) and low energy consumption (0.114 kWh)
  • Mastercycler X50s with large 7 in. touchscreen can control up to 9 eco units
  • Easy program transfer
TRUSTED_SUSTAINABILITY
Specifications
Block Format Fixed
Thermal Accuracy ±0.15°C
Content And Storage Operating Manual, Certificate of Uniformity, and Main Power Cord with EU and US Plugs
Thermal Uniformity Block Homogeneity 20°C to 72°C ≤ ±0.2°C, 95°C ≤ ±0.3°C
Dimensions 27.5 x 43 x 33 cm
Capacity 96
Voltage 110/230 V
Display Type Touchscreen
Product Type Real Time PCR Instrument
Height (English) 13 in.
Depth (English) 17 in.
Height (Metric) 33 cm
Width (Metric) 27.5 cm
Warranty 2 Years
Width (English) 10.8 in.
Depth (Metric) 43 cm
Thermal Range 4°C to 99°C
For Use With (Application) PCR
Length (English) 17 in.
Length (Metric) 43 cm
Wattage 850 W
Max. Ramp Rate 10°C/s
No. of Programs 1000
Gradient Range 1°C to 30°C
Heated Lid 37°C to 110°C
Max. Cooling Rate 5°C/s
Weight 11.5 kg (25.4 lb.)
Block Configurations 96-well