Learn More
Eppendorf™ Mastercycler™ Nexus
Product Code. 12314943 Shop All Eppendorf Products
Change view
Click to view available options
Depth: 410mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12314943 Depth: 410mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12314943 Supplier Eppendorf™ Supplier No. 6333000014
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Voltage 230 V
Electrical Requirements 230 V, 50/60 Hz, 700 W
Product Type Mastercycler Nexus
Height (Metric) 320 mm
Width (Metric) 250 mm
Depth (Metric) 410 mm
Frequency 50 to 60 Hz
Ramp Rate 3°C/s
Material Aluminum