missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22522-0.025mg
This item is not returnable.
View return policy
Description
MAL Polyclonal specifically detects MAL in Human, Mouse, Rat, Xenopus samples. It is validated for Western Blot.
Specifications
| MAL | |
| Polyclonal | |
| Western Blot 1:500 | |
| mal, T-cell differentiation protein, myelin and lymphocyte protein, T-cell differentiation protein MAL, T-lymphocyte maturation-associated protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4118 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS with 1 mg/mL BSA with 0.05% Sodium Azide | |
| MAL | |
| Synthetic peptides corresponding to residues C(114)F D G F T Y (R/K) H Y H E N I(126) of human MAL. | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat, Xenopus species | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction