missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
187.00€
Specifications
| Antigen | MAL |
|---|---|
| Dilution | Western Blot 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MAL Polyclonal specifically detects MAL in Human, Mouse, Rat, Xenopus samples. It is validated for Western Blot.Specifications
| MAL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 1 mg/mL BSA with 0.05% Sodium Azide | |
| 4118 | |
| Synthetic peptides corresponding to residues C(114)F D G F T Y (R/K) H Y H E N I(126) of human MAL. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat, Xenopus species | |
| mal, T-cell differentiation protein, myelin and lymphocyte protein, T-cell differentiation protein MAL, T-lymphocyte maturation-associated protein | |
| MAL | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title