Learn More
Magnesium Standard For ICP, 1000μg/mL in 2% HCl, SPEX CertiPrep™
Product Code. 10325892 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 10325892
Brand: SPEX Certiprep PLMG12X
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
AnalyteConcentration
Mg1000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging Triple-leached LDPE bottle
CAS 7439-95-4, 7647-01-0, 7732-18-5
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
CAS Min % 0.1, 2.0, 97.9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.1, 2.0, 97.9
Molecular Formula Mg
MDL Number MFCD00085308
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [Mg]
IUPAC Name magnesium
Solvent or Matrix 2% HCl
Molecular Weight (g/mol) 24.30
PubChem CID 5462224
Quantity 500 mL
ChEBI CHEBI:25107
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLMG2-2Y, -2T, -2X    10,000    PLMG2-3Y, -3X
2-5% HCl    1000μg/mL    PLMG1-2X
Chemical Name or Material Magnesium Standard
Show More Show Less
Product intended for laboratory use only